missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS OX40 Signaling Assay
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter OX40 Checkpoint Signaling cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting signaling checkpoints. This cell line enables assessment of OX40 induced non-canonical NF-κB signaling via detection of NIK stabilization. This product uses EFC technology. The included cell line overexpresses ED-tagged NIK (NF-kappa-B Inducing Kinase) protein as well as untagged OX40 receptor. Upon activation of the receptor by its ligand or an antibody, NIK is stabilized, allowing detection of EFC signal when treated with exogenous EA. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | non-canonical NF-κB signaling via detection of NIK stabilization |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | OX40L |
| Quantity | 2 Vials |
| Target Entry | OX40 |
| Accession Number | NM_003327.3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction