missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PATJ Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142086
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
This gene encodes a protein with multiple PDZ domains. PDZ domains mediate protein-protein interactions, and proteins with multiple PDZ domains often organize multimeric complexes at the plasma membrane. This protein localizes to tight junctions and to the apical membrane of epithelial cells. A similar protein in Drosophila is a scaffolding protein which tethers several members of a multimeric signaling complex in photoreceptors.
Specifications
| PATJ | |
| Polyclonal | |
| Unconjugated | |
| PATJ | |
| channel-interacting PDZ domain protein; channel-interacting PDZ domain-containing protein; Cipp; hINADL; inactivation no after-potential D-like protein; inactivation-no-afterpotential D-like; Inadl; Inadl protein; Inadl2; InaD-like; InaD-like (Drosophila); InaD-like 2; inaD-like protein; novel ankyrin-repeat protein; PALS1-associated tight junction protein; Patj; PATJ, crumbs cell polarity complex component; PDZ domain protein; protein associated to tight junctions; RGD1565362 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10207 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8NI35 | |
| PATJ | |
| Peptide with sequence PENPATDKLQVLQ-C (aa 2-14). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction