Learn More
PATZ1 Monoclonal Antibody (OTI2H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809257S
This item is not returnable.
View return policy
Description
PATZ1 Monoclonal antibody specifically detects PATZ1 in Human samples. It is validated for Western BlotSpecifications
| PATZ1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q06587 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6015 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2H4 | |
| Unconjugated | |
| RING1 | |
| 8430401L15Rik; AI447519; BTB/POZ domain zinc finger transcription factor; BTB-POZ domain zinc finger transcription factor; dJ400N23; Mazr; MAZ-related factor; Patz; PATZ1; POZ (BTB) and AT hook containing zinc finger 1; POZ-, AT hook-, and zinc finger-containing protein 1; POZ/BTB and AT hook containing zinc finger 1; POZ-AT hook-zinc finger protein; protein kinase A RI subunit alpha-associated protein; RIAZ; transcription factor (MAZ-related) MAZR; ZBTB19; Zfp278; zi; Zinc finger and BTB domain-containing protein 19; zinc finger protein 278; Zinc finger sarcoma gene protein; ZNF278; ZSG | |
| Human recombinant protein fragment corresponding to amino acids 88-362 of human RING1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |