Learn More
PATZ1 Mouse anti-Human, Clone: OTI2H4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809257
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene contains an A-T hook DNA binding motif which usually binds to other DNA binding structures to play an important role in chromatin modeling and transcription regulation. Its Poz domain is thought to function as a site for protein-protein interaction and is required for transcriptional repression, and the zinc-fingers comprise the DNA binding domain. Since the encoded protein has typical features of a transcription factor, it is postulated to be a repressor of gene expression. In small round cell sarcoma, this gene is fused to EWS by a small inversion of 22q, then the hybrid is thought to be translocated (t(1;22)(p36.1;q12). The rearrangement of chromosome 22 involves intron 8 of EWS and exon 1 of this gene creating a chimeric sequence containing the transactivation domain of EWS fused to zinc finger domain of this protein. This is a distinct example of an intra-chromosomal rearrangement of chromosome 22. Four alternatively spliced transcript variants are described for this gene.Specifications
| PATZ1 | |
| Monoclonal | |
| Unconjugated | |
| PATZ1 | |
| MAZR, PATZ, RIAZ, ZBTB19, ZNF278, ZSG, dJ400N23 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23598 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2H4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9HBE1 | |
| PATZ1 | |
| Full length human recombit protein of human PATZ1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.