Learn More
Pavinetant, MedChemExpress
Product Code. 16446068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16446068
Brand: Medchem Express HY14432
MedChemExpress Pavinetant (MLE-4901) is a neurokinin-3 receptor (NK3R) antagonist.
Pavinetant (MLE-4901) is a neurokinin-3 receptor (NK3R) antagonist.
Specifications
Chemical Name or Material Pavinetant
CAS 941690-55-7
Color White
Purity Grade Notes Research
Molecular Formula C26H25N3O3S
Quantity 10 mg
Synonym MLE-4901 AZD2624 AZD4901
Solubility Information DMSO : ≥ 50 mg/mL (108.80 mM)
SMILES CC[C@H](NC(C1=C(C=CC=C2)C2=NC(C3=CC=CC=C3)=C1NS(C)(=O)=O)=O)C4=CC=CC=C4
Molecular Weight (g/mol) 459.56
Formula Weight 459.56
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only