missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAWR/ PAR4 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-219IR
This item is not returnable.
View return policy
Description
PAWR / PAR4 Polyclonal antibody specifically detects PAWR / PAR4 in Human samples. It is validated for Western Blot
Specifications
| PAWR / PAR4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| Par-4, PAR4prostate apoptosis response protein 4, PRKC apoptosis WT1 regulator protein, PRKC, apoptosis, WT1, regulator, Prostate apoptosis response 4 protein, prostate apoptosis response protein PAR-4, transcriptional repressor PAR4, WT1-interacting protein | |
| Synthetic peptides corresponding to the N-terminal sequence (MATGGYRTSSGLGGS) and C-terminal sequence (QENKTLLKVVGQLTR) of human PAR4. | |
| 0.1 mL | |
| Apoptosis, Cancer, GPCR, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| 5074 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction