missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax2 Antibody (PAX2/8671R), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20755AF594
This item is not returnable.
View return policy
Description
Pax2 Monoclonal antibody specifically detects Pax2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Pax2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
| Recombinant fragment (around aa 223-354) of human Pax2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
| 5076 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| PAX2/8671R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction