Learn More
PAX3 Mouse anti-Human, Clone: OTI4D1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809630
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.Specifications
| PAX3 | |
| Monoclonal | |
| Unconjugated | |
| PAX3 | |
| CDHS, HUP2, WS1, WS3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5077 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4D1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P23760 | |
| PAX3 | |
| Full length human recombit protein of human PAX3 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.