Learn More
Invitrogen™ PAX3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142851
Description
Additional Information: This antibody is expected to recognize isoforms PAX3 (NP_852122.1), PAX3d (NP_852123.1), PAX3e (NP_852124.1), PAX3g (NP_852126.1), PAX3h (NP_852125.1), and PAX3i (NP_001120838.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.
Specifications
| PAX3 | |
| Polyclonal | |
| Unconjugated | |
| PAX3 | |
| CDHS; homeodomain protein PAX3; HUP2; MGC120381; MGC120382; MGC120383; MGC120384; MGC134778; paired box 3; paired box gene 3; paired box homeotic gene 3; paired box protein Pax-3; paired domain gene 3; paired domain gene HuP2; PAX3; Pax-3; PAX3/FKHR fusion; Sp; splotch; WS1; WS3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 114502, 18505, 5077 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P23760, P24610 | |
| PAX3 | |
| Peptide with sequence C-STVHQSTIPSNPD (aa 337-349). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.