missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax4 Antibody (OTI2F3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73257PE
This item is not returnable.
View return policy
Description
Pax4 Monoclonal antibody specifically detects Pax4 in Human samples. It is validated for Western Blot
Specifications
| Pax4 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 5078 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot | |
| OTI2F3 | |
| Western Blot | |
| KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4 | |
| Human recombinant protein fragment corresponding to amino acids 1-228 of human PAX4 (NP_006184) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction