missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Pax4 Antibody (PAX4/3011), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26702-100ug
This item is not returnable.
View return policy
Description
Pax4 Monoclonal antibody specifically detects Pax4 in Human samples. It is validated for ELISA
Specifications
| Pax4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5078 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA | |
| PAX4/3011 | |
| Unconjugated | |
| KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4 | |
| Recombinant fragment (around aa1-200) of human Pax4 protein (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction