Learn More
PAX4 Mouse anti-Human, Clone: OTI6H4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF802076
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells.Specifications
| PAX4 | |
| Monoclonal | |
| Unconjugated | |
| PAX4 | |
| KPD, MODY9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5078 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6H4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O43316 | |
| PAX4 | |
| Human recombit protein fragment corresponding to amino acids 1-228 of human PAX4 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.