missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAX4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15547291
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15547291 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15547291 Supplier Invitrogen™ Supplier No. PA1108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-108 has been validated for WB on recombinant human Pax4 protein, but may not be suitable for detecting endogenous Pax4. By immunofluorescence, PA1-108 has been used to stain paraffin-embedded human and mouse pancreatic tissue.

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAX4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene PAX4
Gene Accession No. O43316, P32115
Gene Alias KPD; MGC129960; MODY9; paired box 4; paired box gene 4; paired box protein Pax-4; paired domain gene 4; Pax4; Pax-4
Gene Symbols PAX4
Host Species Rabbit
Immunogen Amino acids 128-350 of human PAX4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18506, 5078
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.