missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596316
This item is not returnable.
View return policy
Description
Immunogen sequence: LGRYYRTGVL EPKGIGGSKP RLATPPVVAR IAQLKGECPA LFAWEIQRQL CAEGLCTQDK TPSVSSINRV LRALQEDQGL PCTRLRSPAV LAPAVLTPHS GSETPRGTHP GTGHRNRTIF SPSQAEALEK EFQRGQYPDS VARGKLATAT S; Positive Samples: Lovo; Cellular Location: Nucleus.
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells.
Specifications
| PAX4 | |
| Polyclonal | |
| Unconjugated | |
| PAX4 | |
| KPD; MGC129960; MODY9; paired box 4; paired box gene 4; paired box protein Pax-4; paired domain gene 4; Pax4; Pax-4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18506, 5078 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43316, P32115 | |
| PAX4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-200 of human PAX4 (NP_0061842). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction