missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax5/BSAP Antibody (PAX5/3977R), DyLight 680, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08310FR
This item is not returnable.
View return policy
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Pax5/BSAP | |
| Monoclonal | |
| DyLight 680 | |
| B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
| A recombinant fragment (around aa 235-382) of human PAX5 protein (exact sequence is proprietary) (Uniprot: Q02548) | |
| 100 μg | |
| B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
| 5079 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PAX5/3977R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction