missing translation for 'onlineSavingsMsg'
Learn More

Pax5/BSAP Mouse anti-Human, Clone: PAX5/2595, Novus Biologicals™

Mouse Monoclonal Antibody

€ 484.00

Specifications

Antigen Pax5/BSAP
Clone PAX5/2595
Concentration 0.2 mg/mL
Dilution ELISA 1-5 μg/mL
Applications ELISA
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18604089
View Documents
Novus Biologicals
NBP3-07580-100ug
100 μg
€ 484.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pax5/BSAP Monoclonal specifically detects Pax5/BSAP in Human samples. It is validated for ELISA.
Specifications

Specifications

Pax5/BSAP
0.2 mg/mL
ELISA
Unconjugated
Mouse
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators
10 mM PBS with 0.05% BSA
5079
IgG1 κ
Protein A or G purified
PAX5/2595
ELISA 1-5 μg/mL
Monoclonal
Purified
RUO
Human
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
Recombinant full-length human Pax5/BSAP protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado