Learn More
Pax6 Antibody (CL5414) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61411G
Product Code. 30289410
  • € 737.00 / 0.10mL
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pax6
Monoclonal
DyLight 488
keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6
This antibody was developed using a recombinant protein derived from P26367, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
5080
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL5414
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat