missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax6 Antibody (PAX6/8578R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 311.00 - € 553.00
Specifications
| Antigen | Pax6 |
|---|---|
| Clone | PAX6/8578R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616354
|
Novus Biologicals
NBP3-20250-20ug |
20 μg |
€ 311.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624724
|
Novus Biologicals
NBP3-20250-100ug |
100 μg |
€ 553.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Pax6 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 10mM PBS with 0.05% BSA | |
| 5080 | |
| IgG κ | |
| Protein A or G purified |
| PAX6/8578R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title