missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax6 Antibody (S03-8G4), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | Pax6 |
|---|---|
| Clone | S03-8G4 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658453
|
Novus Biologicals
NBP3-19729-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626563
|
Novus Biologicals
NBP3-19729-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Pax6 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| A synthetic peptide of human Pax6 (Uniprot # P26367) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S03-8G4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5080 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title