missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX6, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00005080-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MQNSHSGVNQLGGVFVNGRPLPDSTRQKIVELAHSGARPCDISRILQVSNGCVSKILGRYYETGSIRPRAIGGSKPRVATPEVVSKIAQYKRECPSIFAWEIRDRLLSEGVCTNDNIPSVSSINRVLRNLASEKQQMGADGMYDKLRMLNGQTGSWGTRPGWYPGTSVPGQPTQDGCQQQEGGGENTNSISSNGEDSDEAQMRLQLKRKLQRNRTSFTQEQIEALEKEFERTHYPDVFARERLAAKIDLPEARIQVWFSNRRAKWRREEKLRNQRRQASNTPSHIPISSSFSTSVYQPIPQPTTPVSSFTSGSMLGRTDTALTNTYSALPPMPSFTMANNLPMQPPVPSQTSSYSCMLPTSPSVNGRSYDTYTPPHMQTHMNSQPMGTSGTTSTGLISPGVSVPVQVPGSEPDMSQYWPRLQSpecifications
| PAX6 | |
| Polyclonal | |
| No additive | |
| NM_000280.2 | |
| PAX6 | |
| PAX6 (NP_000271.1, 1 a.a. ∼ 422 a.a) full-length human protein. | |
| RUO | |
| 5080 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| paired box 6 | |
| AN/AN2/D11S812E/MGC17209/MGDA/WAGR | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction