missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAX6 Recombinant Rabbit Monoclonal Antibody (HL2599)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556833
This item is not returnable.
View return policy
Description
PAX6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter's anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene.
Specifications
| PAX6 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P26367, P63015, P63016 | |
| PAX6 | |
| Recombinant fragment of Human PAX6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2599 | |
| Unconjugated | |
| PAX6 | |
| 1500038E17Rik; AEY11; AN; AN2; Aniridia; aniridia type II protein; D11S812E; Dey; Dickie's small eye; FVH1; Gsfaey11; Keratitis; MGC17209; MGDA; oculorhombin; paired box 6; paired box gene 6; Paired Box Gene-6; paired box homeotic gene 6; paired box homeotic gene-6; paired box protein Pax-6; Pax6; Pax-6; PAX6 protein; Sey; small eye; transcription factor; WAGR | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18508, 25509, 483441, 5080 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction