missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX7 Monoclonal Antibody (OTI4G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811671
This item is not returnable.
View return policy
Description
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| PAX7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23759 | |
| PAX7 | |
| Human recombit protein fragment corresponding to amino acids 34-491 of human PAX7 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G5 | |
| Unconjugated | |
| PAX7 | |
| HUP1, PAX7B, RMS2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5081 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction