missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX7 Monoclonal Antibody (OTI4G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811671S
This item is not returnable.
View return policy
Description
PAX7 Monoclonal antibody specifically detects PAX7 in Human samples. It is validated for Western BlotSpecifications
| PAX7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z3I7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 137209 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4G5 | |
| Unconjugated | |
| ZNF572 | |
| FLJ37460; HUP1; paired box 7; paired box gene 7; paired box homeotic gene 7; paired box protein Pax-7; paired box transcription factor PAX7; paired domain gene 7; PAX7; Pax-7; PAX7 transcriptional factor; PAX7A; PAX7B; RGD1564360; RMS2 | |
| Full length human recombinant protein of human ZNF572 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction