Learn More
PAX8 Antibody (CL7834) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21161JF669
Product Code. 30308424
  • € 759.00 / 0.10mL
Description
PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PAX8
Monoclonal
Janelia Fluor 669
paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8
This antibody was developed using a synthetic peptide derived from Q06710, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
7849
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL7834
50mM Sodium Borate
Mouse
Protein A purified