missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX8 Antibody (CL7834), Novus Biologicals™
Mouse Monoclonal Antibody
€ 401.00 - € 648.00
Specifications
| Antigen | PAX8 |
|---|---|
| Clone | CL7834 |
| Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687704
|
Novus Biologicals
NBP3-21161-100ul |
100 μg |
€ 648.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689744
|
Novus Biologicals
NBP3-21161-25ul |
25 μg |
€ 401.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PAX8 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
| This antibody was developed against a Synthetic Peptide of Q06710. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL7834 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS, pH 7.2, 40% glycerol | |
| 7849 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title