missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX8 Antibody (ZR-1), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08307AF647
This item is not returnable.
View return policy
Description
PAX8 Monoclonal antibody specifically detects PAX8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PAX8 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
| Synthetic peptide corresponding to the C-terminus of human PAX8 protein (Uniprot: Q06710) | |
| 100 μg | |
| Cancer | |
| 7849 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| ZR-1 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction