missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAX9 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA528044

Product Code. 13412157

  • € 518.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: HeLa, HepG2. Predicted reactivity: Mouse (97%), Rat (96%), Chicken (84%), Rhesus Monkey (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 9 gene is unknown but it may involve development of stratified squamous epithelia as well as various organs and skeletal elements.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAX9
Polyclonal
Unconjugated
pax9
EMBL:BAE79272.1}; etID309776.17; paired box 9; paired box gene 9; paired box homeotic gene 9; paired box protein Pax-9; paired domain gene 9; Pax9; Pax-9; pax9 {ECO:0000312; pax9 protein; Pax9a; Pax9b; STHAG3
Rabbit
Antigen affinity chromatography
RUO
5083
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P55771
pax9
Recombinant protein encompassing a sequence within the center region of human PAX9. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.