missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Paxillin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578143
This item is not returnable.
View return policy
Description
Positive Control: K562, A431, HeLa, HepG2 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Paxillin is a focal adhesion protein and a substrate for several tyrosine kinases such as src, FAK, and p210BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. Paxillin associates tightly with FAK and Crk through its SH2 domain. This interaction is independent of the extracellular matrix. Although paxillin was initially discovered in fibroblasts, its phosphorylation may also be important during neurite extension during differentiation.
Specifications
| Paxillin | |
| Polyclonal | |
| Unconjugated | |
| PXN | |
| adaptor protein; AW108311; AW123232; FLJ16691; hCG_1778014; I79_001229; myocardial ischemic preconditioning associated protein 7; Pax; PAX-1; PAXI; Paxillin; paxillin a; paxillin alpha; pxn; pxna; testicular tissue protein Li 134; TGFB1I1; transforming growth factor beta 1 induced transcript 1; wu:fw71f12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5829 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P49023 | |
| PXN | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Paxillin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction