missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paxillin, Rabbit anti-Human, Clone: LD4 sAB, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 262.00 - € 595.00
Specifications
| Antigen | Paxillin |
|---|---|
| Clone | LD4 sAB |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18380771
|
Novus Biologicals
NBP2-81063-0.025MG |
0.025 mg |
€ 262.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18228109
|
Novus Biologicals
NBP2-81063-0.2MG |
0.2 mg |
€ 595.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Paxillin Monoclonal specifically detects Paxillin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Paxillin | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Phospho Specific, Signal Transduction | |
| PBS | |
| 5829 | |
| Peptide corresponding to human paxillin-LD4 motif (residues 260-278) fused to MBP; peptide sequence: WGGSATRELDELMASLSD. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| LD4 sAB | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ16691, paxillin | |
| PXN | |
| IgG κ | |
| Protein A purified | |
| The antibody binds specifically to the LD-4 motif of human Paxillin with a Kd of 6 nM. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title