missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pb Polyclonal Antibody
GREENER_CHOICE

Product Code. 15847923
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847923 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15847923 Supplier Invitrogen™ Supplier No. PA569513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

For short term use, store at 2-8C up to 1 week. For long term storage, store at -20C in small aliquots to prevent freeze-thaw cycles. Concentration is batch dependent: 0.5-1 mg/mL.

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Controls development of mouthparts, and labial and maxillary palps.
TRUSTED_SUSTAINABILITY

Specifications

Antigen pb
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene pb
Gene Accession No. P31264
Gene Alias BG:DS01719.1; CG17880; CG31481; CG31481-PA; CG31481-PB; CG31481-PC; CG31481-PD; Dmel\CG31481; Dmel_CG31481; Dmpb; Homeotic protein proboscipedia; l(3)04498; PB; pb-PA; pb-PB; pb-PC; pb-PD; pd; pr; proboscipedia; proboscopedia; proposcipedia
Gene Symbols pb
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the middle region of Drosophila pb (aa 236-285).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 40826
Target Species Drosophila
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.