missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBDC1 Antibody (OTI4E11), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71912MFV500
This item is not returnable.
View return policy
Description
PBDC1 Monoclonal antibody specifically detects PBDC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| PBDC1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 51260 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI4E11 | |
| Flow Cytometry | |
| chromosome X open reading frame 26, hypothetical protein LOC51260, MGC874,2610029G23Rik | |
| Full length human recombinant protein of human CXorf26(NP_057584) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction