missing translation for 'onlineSavingsMsg'
Learn More

PBDC1 Antibody (OTI5A11), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-71911AF350

Product Code. 18168342

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PBDC1 Monoclonal specifically detects PBDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PBDC1
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
PBDC1
Full length recombinant protein of human CXorf26 (NP_057584) produced in HEK293T cells.
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI5A11
Western Blot, Immunohistochemistry
chromosome X open reading frame 26, hypothetical protein LOC51260, MGC874,2610029G23Rik
Mouse
0.1 mL
51260
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.