missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PBEF/Visfatin/NAMPT Antibody (Alt-100) - Humanized, Novus Biologicals™

Product Code. 30044733 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044733 100 μg 100µg
30045648 1 mg 1mg
30044483 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30044733 Supplier Novus Biologicals™ Supplier No. NBP328222100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

PBEF/Visfatin/NAMPT Monoclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBEF/Visfatin/NAMPT
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone Alt-100
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. P43490
Gene Alias EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN
Host Species Human
Immunogen NAMPT
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10135
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.