missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBLD Antibody (OTI7G5), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73262JF549
This item is not returnable.
View return policy
Description
PBLD Monoclonal specifically detects PBLD in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| PBLD | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PBLD | |
| Full length recombinant protein of human PBLD (NP_071412) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI7G5 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| EC 5.1, FLJ14767, FLJ35507, MAWBPMAWDBPMAWD binding protein, MAWD-binding protein, phenazine biosynthesis-like domain-containing protein, phenazine biosynthesis-like protein domain containing, Unknown protein 32 from 2D-page of liver tissue | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 64081.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction