missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBR Antibody (CL13358), Novus Biologicals™
Mouse Monoclonal Antibody
€ 429.00 - € 611.00
Specifications
| Antigen | PBR |
|---|---|
| Clone | CL13358 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18633485
|
Novus Biologicals
NBP3-18567-100ul |
100 μg |
€ 611.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602885
|
Novus Biologicals
NBP3-18567-25ul |
25 μg |
€ 429.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PBR Monoclonal antibody specifically detects PBR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PBR | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/ Immunofluorescence 2-10 μg/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| benzodiazepine peripheral binding site, BZRPDBI, IBP, MBRPBS, mDRC, Mitochondrial benzodiazepine receptor, PBRBPBS, PBRS, peripheral benzodiazepine receptor, Peripheral-type benzodiazepine receptor, pk18, PKBSbenzodiazapine receptor (peripheral), PTBR, translocator protein, translocator protein (18kDa) | |
| This antibody was developed against a sythetic peptide for P30536. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL13358 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS, pH 7.2, 40% glycerol | |
| 706 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title