missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PBRM1 Recombinant Rabbit Monoclonal Antibody (BL-39-2C3)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544234
This item is not returnable.
View return policy
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
The PBRM1 gene encodes a subunit of the PBAF (Polybromo-associated BAF) chromatin remodeling complex, playing a crucial role in the regulation of chromatin structure and transcription. PBRM1, also known as BAF180, contains bromodomains that recognize acetylated lysine residues on histone tails, facilitating chromatin remodeling necessary for effective transcriptional regulation. This gene is frequently mutated in renal cell carcinoma, particularly clear cell renal cell carcinoma (ccRCC), where such mutations are implicated in the pathogenesis of the disease. PBRM1 acts as a tumor suppressor, and its loss of function can lead to deregulation of gene expression, contributing to oncogenic processes. It is involved in pathways that govern cell cycle regulation and DNA damage response, reflecting its importance in maintaining genomic stability. Ongoing research focuses on understanding PBRM1's interactions within chromatin remodeling complexes and its potential as a therapeutic target in cancer treatment.
Specifications
| PBRM1 | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
| Q86U86 | |
| Pbrm1 | |
| Recombinant protein representing residues 1-293 of human Polybromo 1. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| BL-39-2C3 | |
| Unconjugated | |
| Pbrm1 | |
| 2310032M22Rik; 2610016F04Rik; AI507524; BAF180; BRG1-associated factor 180; glutamate receptor interacting protein 2; hPB1; LOC506557 protein; PB1; PBRM1; polybromo 1; polybromo-1D; Protein polybromo-1; RGD1565549 | |
| Rabbit | |
| Purified | |
| RUO | |
| 55193 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction