Learn More
PBS, (1X PBS Liq.), w/o Calcium and Magnesium, 500mL, MP Biomedicals
Dulbecco's Formula Without calcium and magnesium Storage temperature: 15-30°C
Brand: MP Biomedicals 091860454
39.24 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : Weight : 0.05000kg
Specifications
500 mL |