missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX1 Antibody (PCRP-PBX1-3C8), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14035MFV500
This item is not returnable.
View return policy
Description
PBX1 Monoclonal antibody specifically detects PBX1 in Human samples. It is validated for Flow Cytometry
Specifications
| PBX1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5087 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| PCRP-PBX1-3C8 | |
| Flow Cytometry | |
| DKFZp686B09108, Homeobox protein PBX1, Homeobox protein PRL, MGC126627, Pre-B cell leukemia transcription factor-1, pre-B-cell leukemia homeobox 1, pre-B-cell leukemia transcription factor 1, PRL | |
| Recombinant full-length human PBX1 protein (Uniprot: P40424) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction