missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBX2 Antibody (PCRP-PBX2-1C4), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14127MFV610
This item is not returnable.
View return policy
Description
PBX2 Monoclonal antibody specifically detects PBX2 in Human samples. It is validated for Flow Cytometry
Specifications
| PBX2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5089 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| PCRP-PBX2-1C4 | |
| Flow Cytometry | |
| G17Homeobox protein PBX2, HOX12homeobox 12, PBX2MHC, pre-B-cell leukemia homeobox 2, pre-B-cell leukemia transcription factor 2, Protein G17 | |
| Recombinant full-length human PBX2 protein (Uniprot: P40425) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction