missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PBX4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA562066

Product Code. 15789861

  • € 572.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AGDAFLTLRT LASLQPPPGG GCLQSQAQGS WQGATPQPAT ASPAGDPGSI NSST Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 63%.

PBX4 encodes a member of the pre-B cell leukemia transcription factor family. These proteins are homeobox proteins that play critical roles in embryonic development and cellular differentiation both as Hox cofactors and through Hox-independent pathways. The encoded protein contains a homeobox DNA-binding domain, but specific functions of the protein have not been determined. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PBX4
Polyclonal
Unconjugated
PBX4
2410015M21Rik; AI429113; AW045266; Edg4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 4; homeobox protein PBX4; PBX homeobox 4; PBX4; pre B cell leukemia homeobox 4; pre-B-cell leukemia homeobox 4; pre-B-cell leukemia transcription factor 4
Rabbit
Antigen affinity chromatography
RUO
80714
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BYU1
PBX4
Recombinant protein corresponding to Human PBX4. Recombinant protein control fragment (Product #RP-101300).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.