missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCA Monoclonal Antibody (OTI1B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810969
This item is not returnable.
View return policy
Description
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o fpropionic acidemia type I (PA-1). PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.Specifications
| PCCA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05165 | |
| PCCA | |
| Full length human recombit protein of human PCCA produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B10 | |
| Unconjugated | |
| PCCA | |
| propionyl-CoA carboxylase alpha subunit | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5095 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction