missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19718
This item is not returnable.
View return policy
Description
PCCB Polyclonal specifically detects PCCB in Human samples. It is validated for Western Blot.
Specifications
| PCCB | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| beta polypeptide, mitochondrial, propionyl CoA carboxylase, beta polypeptide | |
| Rabbit | |
| 58 kDa | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5096 | |
| Human | |
| IgG |
| Western Blot | |
| 0.37mg/mL | |
| Western Blot 1:500-1:3000 | |
| P05166 | |
| PCCB | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PCCB. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (100%), Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction