missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCCB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38150-100ul
This item is not returnable.
View return policy
Description
PCCB Polyclonal antibody specifically detects PCCB in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PCCB | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| beta polypeptide, mitochondrial, propionyl CoA carboxylase, beta polypeptide | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 260-539 of human PCCB (NP_000523.2).,, Sequence:, AFENDVDALCNLRDFFNYLPLSSQDPAPVRECHDPSDRLVPELDTIVPLESTKAYNMVDIIHSVVDEREFFEIMPNYAKNIIVGFARMNGRTVGIVGNQPKVASGCLDINSSVKGARFVRFCDAFNIPLITFVDVPGFLPGTAQEYGGIIRHGAKLLYAFAEATVPKVTVITRKAYGGAYDVMSSKHLCGDTNYAWPTAEIAVMGAKGAVEIIFKGHENVEAAQAEYIEKFANPFPAAVRGFVDDIIQPSSTRARICCDLDVLASKKVQRPWRKHANIPL | |
| 100 μL | |
| Stem Cell Markers | |
| 5096 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction