missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121704
This item is not returnable.
View return policy
Description
PCDH15 Polyclonal Antibody for Western Blot, IHC (P)
PCDH15 is a member of the cadherin superfamily. Family members encode integral membrane proteins that mediate calcium-dependent cell-cell adhesion. PCDH15 consists of a signal peptide, 11 extracellular calcium-binding domains, a transmembrane domain and a unique cytoplasmic domain. It plays an essential role in maintenance of normal retinal and cochlear function. Mutations in this gene have been associated with hearing loss, which is consistent with its location at the Usher syndrome type 1F (USH1F) critical region on chromosome 10.
Specifications
| PCDH15 | |
| Polyclonal | |
| Unconjugated | |
| PCDH15 | |
| Ames waltzer; av; BB078305; cadherin-related family member 15; CDHR15; DFNB23; DKFZp667A1711; ENSMUSG00000046980; Gm9815; nmf19; Pcdh15; protocadherin 15; protocadherin 15 CD2; protocadherin 15 CD3 isoform; protocadherin related 15; protocadherin-15; protocadherin-15-CD2; protocadherin-related 15; RP11-449J3.2; USH1F | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 11994 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q99PJ1 | |
| PCDH15 | |
| Recombinant protein Protocadherin 15. The antigen corresponds to amino acid range 45-270 of the target protein. | |
| 20 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction