missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDH18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553686
This item is not returnable.
View return policy
Description
Immunogen sequence: IHKGDITLVP TINGTLPIRS HHRSSPSSSP TLERGQMGSR QSHNSHQSLN SLVTISSNHV PENFSLELTH ATPAVEQVSQ LLSMLHQGQY QPRPSFRGNK YSRSYR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
Protocadherins comprise the largest group within the cadherin family of calcium-dependent cell-cell adhesion molecules. Protocadherin 18 (PCDH18) was initially identified through screening of human and mouse ESTs using conserved cytoplasmic domain motifs. Tissue screening revealed PCDH18 broad expression in tissues such as brain, liver, heart, kidney, lung, and trachea. PCDH18 expression is also temporally and spatially regulated in the mouse embryonic brain and interacts with Disabled-1, an intracellular adapter protein involved in neuronal migration and cell positioning during mammalian brain development. PCDH18 was also found to be expressed in the developing zebrafish neural tube and central nervous system, lending support to the hypothesis that PCDH18 may play a role during brain development.
Specifications
| PCDH18 | |
| Polyclonal | |
| Unconjugated | |
| PCDH18 | |
| 3110038E07Rik; BB095589; DKFZP434B0923; Kiaa1562; PCDH18; PCDH68L; Protocadherin 18; protocadherin 68-like protein; protocadherin-18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54510 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HCL0 | |
| PCDH18 | |
| Recombinant protein corresponding to Human PCDH18. Recombinant protein control fragment (Product #RP-90578). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction