missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH20 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 200.00 - € 468.00
Specifications
| Antigen | PCDH20 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PCDH20 Polyclonal antibody specifically detects PCDH20 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PCDH20 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 64881 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ22218, PCDH13, protocadherin 13, protocadherin 20, protocadherin-13, protocadherin-20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 730-885 of human PCDH20 (NP_073754.2). ILLLDINDNPPLVLFPQSNMSYLLVLPSTLPGSPVTEVYAVDKDTGMNAVIAYSIIGRRGPRPESFRIDPKTGNITLEEALLQTDYGLHRLLVKVSDHGYPEPLHSTVMVNLFVNDTVSNESYIESLLRKEPEINIEEKEPQISIEPTHRKVESVS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title