missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHA10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543335
This item is not returnable.
View return policy
Description
Peptide sequence: ESRLLDSRFP LEGASDADVG ENALLTYKLS PNEYFVLDII NKKDKDKFPV Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.
Specifications
| PCDHA10 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA10 | |
| cadherin-related neuronal receptor 10; cadherin-related neuronal receptor 3; Cnr3; CNR8; CNRN8; CNRS8; Crnr3; CRNR8; KIAA0345-like 4; ortholog to mouse CNR8; Pcdha10; Pcdha14; PCDH-ALPHA10; PCDH-alpha-10; protocadherin alpha 10; protocadherin alpha 14; protocadherin alpha-10; rCNRv10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56139 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y5I2 | |
| PCDHA10 | |
| Synthetic peptide directed towards the N-terminal of human PCDHA10 (aa 143-192). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction