missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542959
This item is not returnable.
View return policy
Description
Peptide sequence: SGYNAWLSYE LQPETASASI PFRVGLYTGE ISTTRALDET DAPRQRLLVL Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%.
The gene encoding PCDHA4 is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. PCDHA4 is a single-pass type I membrane protein. It contains 6 cadherin domains. PCDHA4 is a potential calcium-dependent cell-adhesion protein. It may be involved in the establishment and maintenance of specific neuronal connections in the brain.
Specifications
| PCDHA4 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA4 | |
| cadherin-related neuronal receptor 1; cadherin-related neuronal receptor 4; Cnr1; CNRN1; Cnrv4; Crnr1; KIAA0345-like 10; MGC138307; MGC142169; ortholog of mouse CNR1; Pcdha4; PCDH-ALPHA4; PCDH-alpha-4; protocadherin alpha 4; protocadherin alpha-4; R75250 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UN74 | |
| PCDHA4 | |
| Synthetic peptide directed towards the C-terminal of human PCDHA4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction