missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560748
This item is not returnable.
View return policy
Description
Immunogen sequence: GEIKVNGELD YEDYNSYEIN IDAMDKSTFP LSGHCKVVVK LLDVNDNTP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 65%.
The gene encoding PCDHA5 is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. PCDHA5 is a single-pass type I membrane protein. It contains 6 cadherin domains. PCDHA5 is a potential calcium-dependent cell-adhesion protein. It may be involved in the establishment and maintenance of specific neuronal connections in the brain.
Specifications
| PCDHA5 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA5 | |
| cadherin-related neuronal receptor 5; cadherin-related neuronal receptor 6; Cnr6; CNRN6; CNRS6; Crnr6; KIAA0345-like 9; ortholog of mouse CNR6; PCDHA5; PCDH-ALPHA5; PCDH-alpha-5; protocadherin alpha 5; protocadherin alpha-5; rCNRv05 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56143 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5H7 | |
| PCDHA5 | |
| Recombinant protein corresponding to Human PCDHA5. Recombinant protein control fragment (Product #RP-96995). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction