missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCDHA9 Monoclonal Antibody (3C1)
GREENER_CHOICE

Product Code. 15518922
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15518922 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518922 Supplier Invitrogen™ Supplier No. MA520826

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: DVSPDIKSKF HMDPLSGAIT VIGHMDFEES RAHKIPVEAV DKGFPPLAGH CTLLVEVVDV NDNAPQLTIK TLSVPVKEDA QLGTVIALIS VIDLDADA.

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCDHA9
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C1
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Pcdha9
Gene Accession No. Q9Y5H5
Gene Alias cadherin-related neuronal receptor 9; KIAA0345; KIAA0345-like 5; Pcdha9; PCDH-ALPHA9; PCDH-alpha-9; protocadherin alpha 9; protocadherin alpha-9; rCNRv09
Gene Symbols Pcdha9
Host Species Mouse
Immunogen PCDHA9 (NP_114063, 284 a.a. ~ 381 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9752
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.